missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SURF2 Full-length ORF (NP_059973.2, 1 a.a. - 256 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006835-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141152

  • 4130.00 NOK / 10µg
Estimated Shipment: 06-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is located in the surfeit gene cluster, a group of very tightly linked genes that do not share sequence similarity. The gene shares a bidirectional promoter with SURF1, which is located on the opposite strand. The function of the encoded protein from this housekeeping gene has not yet been determined. [provided by RefSeq]

Sequence: MSELPGDVRAFLREHPSLRLQTDARKVRCILTGHELPCRLPELQVYTRGKKYQRLVRASPAFDYAEFEPHIVPSTKNPHQLFCKLTLRHINKCPEHVLRHTQGRRYQRALCKYEECQKQGVEYVPACLVHRRRRREDQMDGDGPRPREAFWEPTSSDEGGAASDDSMTDLYPPELFTRKDLGSTEDGDGTDDFLTDKEDEKAKPPREKATDEGRRETTVYRGLVQKRGKKQLGSLKKKFKSHHRKPKSFSSCKQPG
Specifications

Specifications

NP_059973.2
Liquid
6835
SURF2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSELPGDVRAFLREHPSLRLQTDARKVRCILTGHELPCRLPELQVYTRGKKYQRLVRASPAFDYAEFEPHIVPSTKNPHQLFCKLTLRHINKCPEHVLRHTQGRRYQRALCKYEECQKQGVEYVPACLVHRRRRREDQMDGDGPRPREAFWEPTSSDEGGAASDDSMTDLYPPELFTRKDLGSTEDGDGTDDFLTDKEDEKAKPPREKATDEGRRETTVYRGLVQKRGKKQLGSLKKKFKSHHRKPKSFSSCKQPG
RUO
SURF2
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
56kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SURF-2
SURF2
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers