missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SULT2B1 Partial ORF (NP_814444, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006820-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136095

  • 6245.00 NOK / 25µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene sulfates dehydroepiandrosterone but not 4-nitrophenol, a typical substrate for the phenol and estrogen sulfotransferase subfamilies. Two alternatively spliced variants that encode different isoforms have been described. [provided by RefSeq]

Sequence: MDGPAEPQIPGLWDTYEDDISEISQKLPGEYFRYKGVPFPVGLYSLESISLAENTQDVRDDDIFIITYPKSGTTWMIEIICLILKEGDPS
Specifications

Specifications

NP_814444
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MDGPAEPQIPGLWDTYEDDISEISQKLPGEYFRYKGVPFPVGLYSLESISLAENTQDVRDDDIFIITYPKSGTTWMIEIICLILKEGDPS
RUO
SULT2B1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6820
SULT2B1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSST2
SULT2B1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers