Learn More
Abnova™ Human STON1 Full-length ORF (NP_006864.2, 1 a.a. - 735 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011037-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Endocytosis of cell surface proteins is mediated by a complex molecular machinery that assembles on the inner surface of the plasma membrane. This gene encodes one of two human homologs of the Drosophila melanogaster stoned B protein. This protein is related to components of the endocytic machinery and exhibits a modular structure consisting of an N-terminal proline-rich domain, a central region of homology specific to the human stoned B-like proteins, and a C-terminal region homologous to the mu subunits of adaptor protein (AP) complexes. Co-transcription of this gene and the neighboring downstream gene generates a rare transcript (SALF), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product. [provided by RefSeq]
Sequence: MCSTNPGKWVTFDDDPAVQSSQKSKNFPLENQGVCRPNGLKLNLPGLREFPSGSSSTSSTPLSSPIVDFYFSPGPPSNSPLSTPTKDFPGFPGIPKAGTHVLYPIPESSSDSPLAISGGESSLLPTRPTCLSHALLPSDHSCTHPTPKVGLPDEVNPQQAESLGFQSDDLPQFQYFREDCAFSSPFWKDEGSDSHFTLDPPGSKKMFSSRNKEMPIDQKSLNKCSLNYICEKLEHLQSAENQDSLRSLSMHCLCAEENASSFVPHTLFRSQPKSGWSFMLRIPEKKNMMSSRQWGPIFLKVLPGGILQMYYEQGLEKPFKEIQLDPYCRLSEPKVENFSVAGKIHTVKIEHVSYTEKRKYHSKTEVVHEPDIEQMLKLGSTSYHDFLDFLTTVEEELMKLPAVSKPKKNYEEQEISLEIVDNFWGKVTKEGKFVESAVITQIYCLCFVNGNLECFLTLNDLELPKRDESYYEKDSEKKGIDILDYHFHKCVNVQEFEQSRIIKFVPLDACRFELMRFKTLYNGDNLPFSLKSVVVVQGAYVELQAFVNMASLAQRSSYAGSLRSCDNIRIHFPVPSQWIKALWTMNLQRQKSLKAKMNRRACLGSLQELESEPVIQVTVGSAKYESAYQAVVWKIDRLPDKNSSLDHPHCLSYKLELGSDQEIPSDWYPFATVQFSVPDTCASRTEVRSLGVESDVQPQKHVQQRACYNIQVEIEKKWIKIDGEDPDKIGDCITQSpecifications
NP_006864.2 | |
Liquid | |
11037 | |
STON1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MCSTNPGKWVTFDDDPAVQSSQKSKNFPLENQGVCRPNGLKLNLPGLREFPSGSSSTSSTPLSSPIVDFYFSPGPPSNSPLSTPTKDFPGFPGIPKAGTHVLYPIPESSSDSPLAISGGESSLLPTRPTCLSHALLPSDHSCTHPTPKVGLPDEVNPQQAESLGFQSDDLPQFQYFREDCAFSSPFWKDEGSDSHFTLDPPGSKKMFSSRNKEMPIDQKSLNKCSLNYICEKLEHLQSAENQDSLRSLSMHCLCAEENASSFVPHTLFRSQPKSGWSFMLRIPEKKNMMSSRQWGPIFLKVLPGGILQMYYEQGLEKPFKEIQLDPYCRLSEPKVENFSVAGKIHTVKIEHVSYTEKRKYHSKTEVVHEPDIEQMLKLGSTSYHDFLDFLTTVEEELMKLPAVSKPKKNYEEQEISLEIVDNFWGKVTKEGKFVESAVITQIYCLCFVNGNLECFLTLNDLELPKRDESYYEKDSEKKGIDILDYHFHKCVNVQEFEQSRIIKFVPLDACRFELMRFKTLYNGDNLPFSLKSVVVVQGAYVELQAFVNMASLAQRSSYAGSLRSCDNIRIHFPVPSQWIKALWTMNLQRQKSLKAKMNRRACLGSLQELESEPVIQVTVGSAKYESAYQAVVWKIDRLPDKNSSLDHPHCLSYKLELGSDQEIPSDWYPFATVQFSVPDTCASRTEVRSLGVESDVQPQKHVQQRACYNIQVEIEKKWIKIDGEDPDKIGDCITQ | |
RUO | |
STON1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
109.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781K2462/MGC149803/MGC149804/SBLF/STN1/STNB1/stoned-b1 | |
STON1 | |
Yes | |
wheat germ expression system |