missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human STMN2 Full-length ORF (AAH06302.1, 1 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011075-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16196132

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Superior cervical ganglion-10 is a neuronal growth-associated protein that shares significant amino acid sequence similarity with the phosphoprotein stathmin (MIM 151442).[supplied by OMIM]

Sequence: MAKTAMAYKEKMKELSMLSLICSCFYPEPRNINIYTYDDMEVKQINKRASGQAFELILKPPSPISEAPRTLASPKKKDLSLEEIQKKLEAAEERRKSQEAQVLKQLAEKREHEREVLQKALEENNNFSKMAEEKLILKMEQIKENREANLAAIIERLQEKERHAAEVRRNKELQVELSG
Specifications

Specifications

AAH06302.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.43kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SCG10/SCGN10/SGC10
STMN2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
11075
STMN2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAKTAMAYKEKMKELSMLSLICSCFYPEPRNINIYTYDDMEVKQINKRASGQAFELILKPPSPISEAPRTLASPKKKDLSLEEIQKKLEAAEERRKSQEAQVLKQLAEKREHEREVLQKALEENNNFSKMAEEKLILKMEQIKENREANLAAIIERLQEKERHAAEVRRNKELQVELSG
RUO
STMN2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers