Learn More
Abnova™ Human STK19 Partial ORF (NP_004188, 255 a.a. - 364 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008859-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a serine/threonine kinase which localizes predominantly to the nucleus. Its specific function is unknown; it is possible that phosphorylation of this protein is involved in transcriptional regulation. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6 and expresses two transcript variants. [provided by RefSeq]
Sequence: QMTQTFGFRDSEITHLVNAGVLTVRDAGSWWLAVPGAGRFIKYFVKGRQAVLSMVRKAKYRELLLSELLGRRAPVVVRLGLTYHVHDLIGAQLVDCISTTSGTLLRLPETSpecifications
NP_004188 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QMTQTFGFRDSEITHLVNAGVLTVRDAGSWWLAVPGAGRFIKYFVKGRQAVLSMVRKAKYRELLLSELLGRRAPVVVRLGLTYHVHDLIGAQLVDCISTTSGTLLRLPET | |
RUO | |
STK19 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8859 | |
STK19 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D6S60/D6S60E/G11/HLA-RP1/MGC117388/RP1 | |
STK19 | |
Recombinant | |
wheat germ expression system |