missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STAG2 Partial ORF (NP_006594.3, 1130 a.a. - 1231 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010735-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: KRLRPEDSFMSVYPMQTEHHQTPLDYNRRGTSLMEDDEEPIVEDVMMSSEGRIEDLNEGMDFDTMDIDLPPSKNRRERTELKPDFFDPASIMDESVLGVSMFSpecifications
NP_006594.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KRLRPEDSFMSVYPMQTEHHQTPLDYNRRGTSLMEDDEEPIVEDVMMSSEGRIEDLNEGMDFDTMDIDLPPSKNRRERTELKPDFFDPASIMDESVLGVSMF | |
RUO | |
STAG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10735 | |
STAG2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686P168/DKFZp781H1753/FLJ25871/SA-2/SA2/bA517O1.1 | |
STAG2 | |
Recombinant | |
wheat germ expression system |