Learn More
Abnova™ Human STAG1 Partial ORF (AAH64699, 1112 a.a. - 1221 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010274-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the SCC3 family and is expressed in the nucleus. It encodes a component of cohesin, a multisubunit protein complex that provides sister chromatid cohesion along the length of a chromosome from DNA replication through prophase and prometaphase, after which it is dissociated in preparation for segregation during anaphase. [provided by RefSeq]
Sequence: LPAPQLTSTVLRENSRPMGDQIQEPESEHGSEPDFLHNRGLMEEDAEPIFEDVMMSSRSQLEDMNEEFEDTMVIDLPPSRNRRERAELRPDFFDSAAIIEDDSGFGMPMFSpecifications
AAH64699 | |
Liquid | |
10274 | |
STAG1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781D1416/SA1 | |
STAG1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LPAPQLTSTVLRENSRPMGDQIQEPESEHGSEPDFLHNRGLMEEDAEPIFEDVMMSSRSQLEDMNEEFEDTMVIDLPPSRNRRERAELRPDFFDSAAIIEDDSGFGMPMF | |
RUO | |
STAG1 | |
Wheat Germ (in vitro) | |
GST |