missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ST3GAL5 Full-length ORF (AAH65936.1, 1 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008869-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193012

  • 4115.00 NOK / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ganglioside GM3 is known to participate in the induction of cell differentiation, modulation of cell proliferation, maintenance of fibroblast morphology, signal transduction, and integrin-mediated cell adhesion. The protein encoded by this gene is a type II membrane protein which catalyzes the formation of GM3 using lactosylceramide as the substrate. The encoded protein is a member of glycosyltransferase family 29 and may be localized to the Golgi apparatus. Mutation in this gene has been associated with Amish infantile epilepsy syndrome. Transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MPSEYTYVKLRSDCSRPSLQWYTRAQSKMRRPSLLLKDILKCTLLVFGVWILYILKLNYTTEECDMKKMHYVDPDRVKRAQKYAQQVLQKECRPKFAKTSMALLFEHRYSVDLLPFVQKAPKDSEAESKYDPPFGFRKFSSKVQTLLELLPEHDLPEHLKAKTCRRCVVIGSGGILHGLELGHTLNQFDVVIRLNSAPVEGYSEHVGNKTTIRMTYPEGAPLSDLEYYSNDLFVAVLFKSVDFNWLQAMVKKETLPFWVRLFFWKQVAEKIPLQPKHFRILNPVIIKETAFDILQYSEPQSRFWGRDKNVPTIGVIAVVLATHLCDEVSLAGFGYDLNQPRTPLHYFDSQCMAAMNFQTMHNVTTETKFLLKLVKEGVVKDLSGGIDREF
Specifications

Specifications

AAH65936.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SIAT9/SIATGM3S/ST3GalV
ST3GAL5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8869
ST3GAL5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPSEYTYVKLRSDCSRPSLQWYTRAQSKMRRPSLLLKDILKCTLLVFGVWILYILKLNYTTEECDMKKMHYVDPDRVKRAQKYAQQVLQKECRPKFAKTSMALLFEHRYSVDLLPFVQKAPKDSEAESKYDPPFGFRKFSSKVQTLLELLPEHDLPEHLKAKTCRRCVVIGSGGILHGLELGHTLNQFDVVIRLNSAPVEGYSEHVGNKTTIRMTYPEGAPLSDLEYYSNDLFVAVLFKSVDFNWLQAMVKKETLPFWVRLFFWKQVAEKIPLQPKHFRILNPVIIKETAFDILQYSEPQSRFWGRDKNVPTIGVIAVVLATHLCDEVSLAGFGYDLNQPRTPLHYFDSQCMAAMNFQTMHNVTTETKFLLKLVKEGVVKDLSGGIDREF
RUO
ST3GAL5
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers