Learn More
Abnova™ Human SREBF2 Partial ORF (AAH56158, 801 a.a. - 900 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006721-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain. [provided by RefSeq]
Sequence: QAFCKNLLERAIESLVKPQAKKKAGDQEEESCEFSSALEYLKLLHSFVDSVGVMSPPLSRSSVLKSALGPDIICRWWTSAITVAISWLQGDDAAVRSHFTSpecifications
AAH56158 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QAFCKNLLERAIESLVKPQAKKKAGDQEEESCEFSSALEYLKLLHSFVDSVGVMSPPLSRSSVLKSALGPDIICRWWTSAITVAISWLQGDDAAVRSHFT | |
RUO | |
SREBF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6721 | |
SREBF2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SREBP2/bHLHd2 | |
SREBF2 | |
Recombinant | |
wheat germ expression system |