missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SPRR2F Full-length ORF (NP_001014450.1, 1 a.a. - 72 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006705-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MSYQQQQCKQPCQPPPVCPAPKCPEPCPPPKCPEPCPPSKCPQSCPPQQCQQKCPPVTPSPPCQPKCPPKSKSpecifications
NP_001014450.1 | |
Liquid | |
6705 | |
SPRR2F (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSYQQQQCKQPCQPPPVCPAPKCPEPCPPPKCPEPCPPSKCPQSCPPQQCQQKCPPVTPSPPCQPKCPPKSK | |
RUO | |
SPRR2F | |
Yes | |
wheat germ expression system |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SPRR2F | |
Wheat Germ (in vitro) | |
GST |