Learn More
Abnova™ Human SOX13 Partial ORF (NP_005677, 25 a.a. - 138 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009580-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. It has also been determined to be a type-1 diabetes autoantigen, also known as islet cell antibody 12. [provided by RefSeq]
Sequence: KSEEKKEPCHEAPQGSATAAEPQPGDPARASQDSADPQAPAQGNFRGSWDCSSPEGNGSPEPKRPGVSEAASGSQEKLDFNRNLKEVVPAIEKLLSSDWKERFLGRNSMEAKDVSpecifications
NP_005677 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.28kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KSEEKKEPCHEAPQGSATAAEPQPGDPARASQDSADPQAPAQGNFRGSWDCSSPEGNGSPEPKRPGVSEAASGSQEKLDFNRNLKEVVPAIEKLLSSDWKERFLGRNSMEAKDV | |
RUO | |
SOX13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9580 | |
SOX13 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ICA12/MGC117216/Sox-13 | |
SOX13 | |
Recombinant | |
wheat germ expression system |