Learn More
Abnova™ Human SNX24 Full-length ORF (AAH10886, 1 a.a. - 169 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand:  Abnova™ H00028966-P01.10ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16128892
  • 4115.00 NOK / 10µg
Estimated Shipment: 28-06-2024
Description
Sequence: MEVYIPSFRYEESDLERGYTVFKIEVLMNGRKHFVEKRYSEFHALHKKLKKCIKTPEIPSKHVRNWVPKVLEQRRQGLETYLQAVILENEELPKLFLDFLNVRHLPSLPKAESCGSFDETESEESSKLSHQPVLLFLGDPYVLPAASDFPNVVIEGVLHGIFYPHLQPR
Specifications
Specifications
AAH10886
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.33kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PRO1284/SBBI31
SNX24
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
28966
SNX24 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEVYIPSFRYEESDLERGYTVFKIEVLMNGRKHFVEKRYSEFHALHKKLKKCIKTPEIPSKHVRNWVPKVLEQRRQGLETYLQAVILENEELPKLFLDFLNVRHLPSLPKAESCGSFDETESEESSKLSHQPVLLFLGDPYVLPAASDFPNVVIEGVLHGIFYPHLQPR
RUO
SNX24