missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SNRPC Full-length ORF (NP_003084.1, 1 a.a. - 159 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006631-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140882

  • 5420.23 NOK / 25µg

Estimated Shipment: 10-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MPKFYCDYCDTYLTHDSPSVRKTHCSGRKHKENVKDYYQKWMEEQAQSLIDKTTAAFQQGKIPPTPFSAPPPAGAMIPPPPSLPGPPRPGMMPAPHMGGPPMMPMMGPPPPGMMPVGPAPGMRPPMGGHMPMMPGPPMMRPPARPMMVPTRPGMTRPDR
Specifications

Specifications

NP_003084.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
43.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20302
SNRPC
Yes
wheat germ expression system
Liquid
6631
SNRPC (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPKFYCDYCDTYLTHDSPSVRKTHCSGRKHKENVKDYYQKWMEEQAQSLIDKTTAAFQQGKIPPTPFSAPPPAGAMIPPPPSLPGPPRPGMMPAPHMGGPPMMPMMGPPPPGMMPVGPAPGMRPPMGGHMPMMPGPPMMRPPARPMMVPTRPGMTRPDR
RUO
SNRPC
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates