missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SMG5 Full-length ORF (NP_056142.1, 1 a.a. - 1016 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023381-P01.25ug
Additional Details : Weight : 0.00010kg
Description
SMG5 is involved in nonsense-mediated mRNA decay (Ohnishi et al., 2003 [PubMed 14636577]).[supplied by OMIM]
Sequence: MSQGPPTGESSEPEAKVLHTKRLYRAVVEAVHRLDLILCNKTAYQEVFKPENISLRNKLRELCVKLMFLHPVDYGRKAEELLWRKVYYEVIQLIKTNKKHIHSRSTLECAYRTHLVAGIGFYQHLLLYIQSHYQLELQCCIDWTHVTDPLIGCKKPVSASGKEMDWAQMACHRCLVYLGDLSRYQNELAGVDTELLAERFYYQALSVAPQIGMPFNQLGTLAGSKYYNVEAMYCYLRCIQSEVSFEGAYGNLKRLYDKAAKMYHQLKKCETRKLSPGKKRCKDIKRLLVNFMYLQSLLQPKSSSVDSELTSLCQSVLEDFNLCLFYLPSSPNLSLASEDEEEYESGYAFLPDLLIFQMVIICLMCVHSLERAGSKQYSAAIAFTLALFSHLVNHVNIRLQAELEEGENPVPAFQSDGTDEPESKEPVEKEEEPDPEPPPVTPQVGEGRKSRKFSRLSCLRRRRHPPKVGDDSDLSEGFESDSSHDSARASEGSDSGSDKSLEGGGTAFDAETDSEMNSQESRSDLEDMEEEEGTRSPTLEPPRGRSEAPDSLNGPLGPSEASIASNLQAMSTQMFQTKRCFRLAPTFSNLLLQPTTNPHTSASHRPCVNGDVDKPSEPASEEGSESEGSESSGRSCRNERSIQEKLQVLMAEGLLPAVKVFLDWLRTNPDLIIVCAQSSQSLWNRLSVLLNLLPAAGELQESGLALCPEVQDLLEGCELPDLPSSLLLPEDMALRNLPPLRAAHRRFNFDTDRPLLSTLEESVVRICCIRSFGHFIARLQGSILQFNPEVGIFVSIAQSEQESLLQQAQAQFRMAQEEARRNRLMRDMAQLRLQLEVSQLEGSLQQPKAQSAMSPYLVPDTQALCHHLPVIRQLATSGRFIVIIPRTVIDGLDLLKKEHPGARDGIRYLEAEFKKGNRYIRCQKEVGKSFERHKLKRQDADAWTLYKILDSCKQLTLAQGAGEEDPSGMVTIITGLPLDNPSVLSGPMQAALQAAAHASVDIKDVLDFYKQWKEIGSpecifications
NP_056142.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
140.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EST1B/FLJ34864/KIAA1089/LPTS-RP1/LPTSRP1/RP11-54H19.7/SMG-5 | |
SMG5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23381 | |
SMG5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSQGPPTGESSEPEAKVLHTKRLYRAVVEAVHRLDLILCNKTAYQEVFKPENISLRNKLRELCVKLMFLHPVDYGRKAEELLWRKVYYEVIQLIKTNKKHIHSRSTLECAYRTHLVAGIGFYQHLLLYIQSHYQLELQCCIDWTHVTDPLIGCKKPVSASGKEMDWAQMACHRCLVYLGDLSRYQNELAGVDTELLAERFYYQALSVAPQIGMPFNQLGTLAGSKYYNVEAMYCYLRCIQSEVSFEGAYGNLKRLYDKAAKMYHQLKKCETRKLSPGKKRCKDIKRLLVNFMYLQSLLQPKSSSVDSELTSLCQSVLEDFNLCLFYLPSSPNLSLASEDEEEYESGYAFLPDLLIFQMVIICLMCVHSLERAGSKQYSAAIAFTLALFSHLVNHVNIRLQAELEEGENPVPAFQSDGTDEPESKEPVEKEEEPDPEPPPVTPQVGEGRKSRKFSRLSCLRRRRHPPKVGDDSDLSEGFESDSSHDSARASEGSDSGSDKSLEGGGTAFDAETDSEMNSQESRSDLEDMEEEEGTRSPTLEPPRGRSEAPDSLNGPLGPSEASIASNLQAMSTQMFQTKRCFRLAPTFSNLLLQPTTNPHTSASHRPCVNGDVDKPSEPASEEGSESEGSESSGRSCRNERSIQEKLQVLMAEGLLPAVKVFLDWLRTNPDLIIVCAQSSQSLWNRLSVLLNLLPAAGELQESGLALCPEVQDLLEGCELPDLPSSLLLPEDMALRNLPPLRAAHRRFNFDTDRPLLSTLEESVVRICCIRSFGHFIARLQGSILQFNPEVGIFVSIAQSEQESLLQQAQAQFRMAQEEARRNRLMRDMAQLRLQLEVSQLEGSLQQPKAQSAMSPYLVPDTQALCHHLPVIRQLATSGRFIVIIPRTVIDGLDLLKKEHPGARDGIRYLEAEFKKGNRYIRCQKEVGKSFERHKLKRQDADAWTLYKILDSCKQLTLAQGAGEEDPSGMVTIITGLPLDNPSVLSGPMQAALQAAAHASV | |
RUO | |
SMG5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |