Learn More
Abnova™ Human SLC38A4 Partial ORF (NP_060488, 245 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055089-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
SLC38A4 is found predominantly in liver and transports both cationic and neutral amino acids. The transport of cationic amino acids by SLC38A4 is Na(+) and pH independent, while the transport of neutral amino acids is Na(+) and pH dependent (Hatanaka et al., 2001 [PubMed 11342143]).[supplied by OMIM]
Sequence: FQIPCPLPVLDHSVGNLSFNNTLPMHVVMLPNNSESSDVNFMMDYTHRNPAGLDENQAKGSLHDSGVEYEAHSDDKCEPKYFVFNSRSpecifications
NP_060488 | |
Liquid | |
55089 | |
SLC38A4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATA3/FLJ10191/MGC126876/NAT3/PAAT | |
SLC38A4 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.31kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FQIPCPLPVLDHSVGNLSFNNTLPMHVVMLPNNSESSDVNFMMDYTHRNPAGLDENQAKGSLHDSGVEYEAHSDDKCEPKYFVFNSR | |
RUO | |
SLC38A4 | |
Wheat Germ (in vitro) | |
GST |