missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC22A1LS Partial ORF (NP_009036, 144 a.a. - 253 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005003-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16142235

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: EVVYSVPDNVPGQNGSRRPLVCKITGKCLSVCSEENAKAGGCSAFPLLLSQLGARMTGREHAHKGPELTTPDSGLPRPPNPALAGFRALAQHSPPLGTSTPSAVLLSAAT
Specifications

Specifications

NP_009036
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EVVYSVPDNVPGQNGSRRPLVCKITGKCLSVCSEENAKAGGCSAFPLLLSQLGARMTGREHAHKGPELTTPDSGLPRPPNPALAGFRALAQHSPPLGTSTPSAVLLSAAT
RUO
SLC22A18AS
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5003
SLC22A1LS (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BWR1B/BWSCR1B/ORCTL2S/SLC22A1LS/p27-BWR1B
SLC22A18AS
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers