Learn More
Abnova™ Human SLC12A6 Partial ORF (NP_005126, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009990-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the K-Cl cotransporter (KCC) family. K-Cl cotransporters are integral membrane proteins that lower intracellular chloride concentrations below the electrochemical equilibrium potential. The proteins encoded by this gene are activated by cell swelling induced by hypotonic conditions. Alternate splicing results in multiple transcript variants encoding different isoforms. Mutations in this gene are associated with agenesis of the corpus callosum with peripheral neuropathy. [provided by RefSeq]
Sequence: MPHFTVTKVEDPEEGAAASISQEPSLADIKARIQDSDEPDLSQNSITGEHSQLLDDGHKKARNAYLNNSNYEEGDEYFDKNLALFEEEMDSpecifications
NP_005126 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPHFTVTKVEDPEEGAAASISQEPSLADIKARIQDSDEPDLSQNSITGEHSQLLDDGHKKARNAYLNNSNYEEGDEYFDKNLALFEEEMD | |
RUO | |
SLC12A6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9990 | |
SLC12A6 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACCPN/DKFZp434D2135/KCC3/KCC3A/KCC3B | |
SLC12A6 | |
Recombinant | |
wheat germ expression system |