missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SIN3A Partial ORF (NP_056292.1, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00025942-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135226

  • 6245.00 NOK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a transcriptional regulatory protein. It contains paired amphipathic helix (PAH) domains, which are important for protein-protein interactions and may mediate repression by the Mad-Max complex. [provided by RefSeq]

Sequence: MKRRLDDQESPVYAAQQRRIPGSTEAFPHQHRVLAPAPPVYEAVSETMQSATGIQYSVTPSYQVSAMPQSSGSHGPAIAAVHSSHHHPTAVQPHGGQVV
Specifications

Specifications

NP_056292.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MKRRLDDQESPVYAAQQRRIPGSTEAFPHQHRVLAPAPPVYEAVSETMQSATGIQYSVTPSYQVSAMPQSSGSHGPAIAAVHSSHHHPTAVQPHGGQVV
RUO
SIN3A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
25942
SIN3A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434K2235/FLJ90319/KIAA0700
SIN3A
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers