Learn More
Abnova™ Human SIGLEC10 Partial ORF (NP_149121, 589 a.a. - 696 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00089790-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6 (MIM 604405).[supplied by OMIM]
Sequence: SRHSTILDYINVVPTAGPLAQKRNQKATPNSPRTPLPPGAPSPESKKNQKKQYQLPSFPEPKSSTQAPESQESQEELHYATLNFPGVRPRPEARMPKGTQADYAEVKFSpecifications
NP_149121 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SRHSTILDYINVVPTAGPLAQKRNQKATPNSPRTPLPPGAPSPESKKNQKKQYQLPSFPEPKSSTQAPESQESQEELHYATLNFPGVRPRPEARMPKGTQADYAEVKF | |
RUO | |
SIGLEC10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
89790 | |
SIGLEC10 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126774/PRO940/SIGLEC-10/SLG2 | |
SIGLEC10 | |
Recombinant | |
wheat germ expression system |