missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEZ6L2 Full-length ORF (AAH00567.1, 1 a.a. - 809 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026470-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16128332

  • 5540.00 NOK / 25µg
Estimated Shipment: 22-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MGTPRAQHPPPPQLLFLILLSCPWIQGLPLKEEEILPEPGSETPTVASEALAELLHGALLRRGPEMGYLPGSDPDPTLATPPAGQTLAVPSLPRATEPGTGPLTTAVTPNGVRGAGPTAPELLTPPPGTTAPPPPSPASPGPPLGPEGGEEETTTTIITTTTVTTTVTSPAYLLSCGFPPRPAHGDVSVTDLHPGGTATFHCDSGYQLQGEETLICLNGTRPSWNGETPSCMASCGGTIHNATLGRIVSPEPGGAVGPNLTCRWVIEAAEGRRLHLHFERVSLDEDNDRLMVRSGGSPLSPVIYDSDMDDVPERGLISDAQSLYVELLSETPANPLLLSLRFEAFEEDRCFAPFLAHGNVTTTDPEYRPGALATFSCLPGYALEPPGPPNAIECVDPTEPHWNDTEPACKAMCGGELSEPAGVVLSPDWPQSYSPGQDCVWGVHVQEEKRILLQVEILNVREGDMLTLFDGDGPSARVLAQLRGPQPRRRLLSSGPDLTLQFQAPPGPPNPGLGQGFVLHFKEVPRNDTCPELPPPEWGWRTASHGDLIRGTVLTYQCEPGYELLGSDILTCQWDLSWSAAPPACQKIMTCADPGEIANGHRTASDAGFPVGSHVQYRCLPGYSLEGAAMLTCYSRDTGTPKWSDRVPKCALKYEPCLNPGVPENGYQTLYKHHYQAGESLRFFCYEGFELIGEVTITCVPGHPSQWTSQPPLCKVAYEELLDNRKLEVTQTTDPSRQLEGGNLALAILLPLGLVIVLGSGVYIYYTKLQGKSLFGFSGSHSYSPITVESDFSNPLYEAGDTREYEVSI
Specifications

Specifications

AAH00567.1
Liquid
26470
SEZ6L2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGTPRAQHPPPPQLLFLILLSCPWIQGLPLKEEEILPEPGSETPTVASEALAELLHGALLRRGPEMGYLPGSDPDPTLATPPAGQTLAVPSLPRATEPGTGPLTTAVTPNGVRGAGPTAPELLTPPPGTTAPPPPSPASPGPPLGPEGGEEETTTTIITTTTVTTTVTSPAYLLSCGFPPRPAHGDVSVTDLHPGGTATFHCDSGYQLQGEETLICLNGTRPSWNGETPSCMASCGGTIHNATLGRIVSPEPGGAVGPNLTCRWVIEAAEGRRLHLHFERVSLDEDNDRLMVRSGGSPLSPVIYDSDMDDVPERGLISDAQSLYVELLSETPANPLLLSLRFEAFEEDRCFAPFLAHGNVTTTDPEYRPGALATFSCLPGYALEPPGPPNAIECVDPTEPHWNDTEPACKAMCGGELSEPAGVVLSPDWPQSYSPGQDCVWGVHVQEEKRILLQVEILNVREGDMLTLFDGDGPSARVLAQLRGPQPRRRLLSSGPDLTLQFQAPPGPPNPGLGQGFVLHFKEVPRNDTCPELPPPEWGWRTASHGDLIRGTVLTYQCEPGYELLGSDILTCQWDLSWSAAPPACQKIMTCADPGEIANGHRTASDAGFPVGSHVQYRCLPGYSLEGAAMLTCYSRDTGTPKWSDRVPKCALKYEPCLNPGVPENGYQTLYKHHYQAGESLRFFCYEGFELIGEVTITCVPGHPSQWTSQPPLCKVAYEELLDNRKLEVTQTTDPSRQLEGGNLALAILLPLGLVIVLGSGVYIYYTKLQGKSLFGFSGSHSYSPITVESDFSNPLYEAGDTREYEVSI
RUO
SEZ6L2
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
113.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ90517/PSK-1
SEZ6L2
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers