missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Albumin Antibody, R&D Systems™

Mouse Monoclonal Antibody
1885.00 NOK - 4720.00 NOK
Specifications
| Antigen | Albumin |
|---|---|
| Clone | 188815 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18782677
![]() |
R&D Systems
MAB14551-SP |
25 μg |
1885.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700657
![]() |
R&D Systems
MAB14551-100 |
100 μg |
4720.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Albumin Monoclonal specifically detects Albumin in Human samples. It is validated for ELISA.Specifications
| Albumin | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| 213 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 188815 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| ALB, albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Human Serum Albumin | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Human Serum Albumin in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title