Learn More
Abnova™ Human SEMA6C Partial ORF (NP_112175, 28 a.a. - 126 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010500-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene product is a member of the semaphorin family of proteins. Semaphorins represent important molecular signals controlling multiple aspects of the cellular response that follows CNS injury, and thus may play an important role in neural regeneration. [provided by RefSeq]
Sequence: QDPLPLLISDLQGTSPLSWFRGLEDDAVAAELGLDFQRFLTLNRTLLVAARDHVFSFDLQAEEEGEGLVPNKYLTWRSQDVENCAVRGKLTDECYNYIRSpecifications
NP_112175 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QDPLPLLISDLQGTSPLSWFRGLEDDAVAAELGLDFQRFLTLNRTLLVAARDHVFSFDLQAEEEGEGLVPNKYLTWRSQDVENCAVRGKLTDECYNYIR | |
RUO | |
SEMA6C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10500 | |
SEMA6C (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SEMAY/m-SemaY/m-SemaY2 | |
SEMA6C | |
Recombinant | |
wheat germ expression system |