missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEMA3D Partial ORF (NP_689967, 674 a.a. - 777 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00223117-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16142527

  • 6270.00 NOK / 25µg
Estimated Shipment: 01-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: HTIVKLTLNVIENEQMENTQRAEHEEGKVKDLLAESRLRYKDYIQILSSPNFSLDQYCEQMWHREKRRQRNKGGPKWKHMQEMKKKRNRRHHRDLDELPRAVAT
Specifications

Specifications

NP_689967
Liquid
223117
SEMA3D (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC39708/Sema-Z2/coll-2
SEMA3D
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.18kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HTIVKLTLNVIENEQMENTQRAEHEEGKVKDLLAESRLRYKDYIQILSSPNFSLDQYCEQMWHREKRRQRNKGGPKWKHMQEMKKKRNRRHHRDLDELPRAVAT
RUO
SEMA3D
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers