missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEC24C Full-length ORF (AAH18928, 1 a.a. - 1094 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009632-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123952

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The product of this gene may play a role in shaping the vesicle, as well as in cargo selection and concentration. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq]

Sequence: MNVNQSVPPVPPFGQPQPIYPGYHQSSYGGQSGSTAPAIPYGAYNGPVPGYQQTPPQGMSRAPPSSGAPPASTAQAPCGQAAYGQFGQGDVQNGPSSTVQMQRLPGSQSFGSPLAPVGNQPPVLQPYGPPPTSAQVATQLSGMQISGAVAPAPPSSGLGFGPPTSLASASGSFPNSGLYGSYPQGQAPPLSQAQGHPGIQTPQRSAPSQASSFTPPASGGPRLPSMTGPLLPGQSFGGPSVSQPNHVSSPPQALPPGTQMTGPLGPLPPMHSPQQPGYQPQQNGSFGPARGPQSNYGGPYPAAPTFGSQPGPPQPLPPKRLDPDAIPSPIQVIEDDRNNRGTEPFVTGVRGQVPPLVTTNFLVKDQGNASPRYIRCTSYNIPCTSDMAKQAQVPLAAVIKPLARLPPEEASPYVVDHGESGPLRCNRCKAYMCPFMQFIEGGRRFQCCFCSCINDVPPQYFQHLDHTGKRVDAYDRPELSLGSYEFLATVDYCKNNKFPSPPAFIFMIDVSYNAIRTGLVRLLCEELKSLLDFLPREGGAEESAIRVGFVTYNKVLHFYNVKSSLAQPQMMVVSDVADMFVPLLDGFLVNVNESRAVITSLLDQIPEMFADTRETETVFVPVIQAGMEALKAAECAGKLFLFHTSLPIAEAPGKLKNRDDRKLINTDKEKTLFQPQTGAYQTLAKECVAQGCCVDLFLFPNQYVDVATLSVVPQLTGGSVYKYASFQVENDQERFLSDLRRDVQKVVGFDAVMRVRTSTGIRAVDFFGAFYMSNTTDVELAGLDGDKTVTVEFKHDDRLNEESGALLQCALLYTSCAGQRRLRIHNLALNCCTQLADLYRNCETDTLINYMAKFAYRGVLNSPVKAVRDTLITQCAQILACYRKNCASPSSAGQLILPECMKLLPVYLNCVLKSDVLQPGAEVTTDDRAYVRQLVTSMDVTETNVFFYPRLLPLTKSPVESTTEPPAVRASEERLSNGDIYLLENGLNLFLWVGASVQQGVVQSLFSVSSFSQITSGLSVLPVLDNPLSKKVRGLIDSLRAQRSRYMKLTVVKQEDKMEMLFKHFLVEDKSLSGGASYVDFLCHMHKEIRQLLS
Specifications

Specifications

AAH18928
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
145.86kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0079
SEC24C
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9632
SEC24C (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNVNQSVPPVPPFGQPQPIYPGYHQSSYGGQSGSTAPAIPYGAYNGPVPGYQQTPPQGMSRAPPSSGAPPASTAQAPCGQAAYGQFGQGDVQNGPSSTVQMQRLPGSQSFGSPLAPVGNQPPVLQPYGPPPTSAQVATQLSGMQISGAVAPAPPSSGLGFGPPTSLASASGSFPNSGLYGSYPQGQAPPLSQAQGHPGIQTPQRSAPSQASSFTPPASGGPRLPSMTGPLLPGQSFGGPSVSQPNHVSSPPQALPPGTQMTGPLGPLPPMHSPQQPGYQPQQNGSFGPARGPQSNYGGPYPAAPTFGSQPGPPQPLPPKRLDPDAIPSPIQVIEDDRNNRGTEPFVTGVRGQVPPLVTTNFLVKDQGNASPRYIRCTSYNIPCTSDMAKQAQVPLAAVIKPLARLPPEEASPYVVDHGESGPLRCNRCKAYMCPFMQFIEGGRRFQCCFCSCINDVPPQYFQHLDHTGKRVDAYDRPELSLGSYEFLATVDYCKNNKFPSPPAFIFMIDVSYNAIRTGLVRLLCEELKSLLDFLPREGGAEESAIRVGFVTYNKVLHFYNVKSSLAQPQMMVVSDVADMFVPLLDGFLVNVNESRAVITSLLDQIPEMFADTRETETVFVPVIQAGMEALKAAECAGKLFLFHTSLPIAEAPGKLKNRDDRKLINTDKEKTLFQPQTGAYQTLAKECVAQGCCVDLFLFPNQYVDVATLSVVPQLTGGSVYKYASFQVENDQERFLSDLRRDVQKVVGFDAVMRVRTSTGIRAVDFFGAFYMSNTTDVELAGLDGDKTVTVEFKHDDRLNEESGALLQCALLYTSCAGQRRLRIHNLALNCCTQLADLYRNCETDTLINYMAKFAYRGVLNSPVKAVRDTLITQCAQILACYRKNCASPSSAGQLILPECMKLLPVYLNCVLKSDVLQPGAEVTTDDRAYVRQLVTSMDVTETNVFFYPRLLPLTKSPVESTTEPPAVRASEERLSNGDIYLLENGLNLFLWVGASVQQG
RUO
SEC24C
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers