Learn More
Abnova™ Human SEC24C Full-length ORF (AAH18928, 1 a.a. - 1094 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009632-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The product of this gene may play a role in shaping the vesicle, as well as in cargo selection and concentration. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq]
Sequence: MNVNQSVPPVPPFGQPQPIYPGYHQSSYGGQSGSTAPAIPYGAYNGPVPGYQQTPPQGMSRAPPSSGAPPASTAQAPCGQAAYGQFGQGDVQNGPSSTVQMQRLPGSQSFGSPLAPVGNQPPVLQPYGPPPTSAQVATQLSGMQISGAVAPAPPSSGLGFGPPTSLASASGSFPNSGLYGSYPQGQAPPLSQAQGHPGIQTPQRSAPSQASSFTPPASGGPRLPSMTGPLLPGQSFGGPSVSQPNHVSSPPQALPPGTQMTGPLGPLPPMHSPQQPGYQPQQNGSFGPARGPQSNYGGPYPAAPTFGSQPGPPQPLPPKRLDPDAIPSPIQVIEDDRNNRGTEPFVTGVRGQVPPLVTTNFLVKDQGNASPRYIRCTSYNIPCTSDMAKQAQVPLAAVIKPLARLPPEEASPYVVDHGESGPLRCNRCKAYMCPFMQFIEGGRRFQCCFCSCINDVPPQYFQHLDHTGKRVDAYDRPELSLGSYEFLATVDYCKNNKFPSPPAFIFMIDVSYNAIRTGLVRLLCEELKSLLDFLPREGGAEESAIRVGFVTYNKVLHFYNVKSSLAQPQMMVVSDVADMFVPLLDGFLVNVNESRAVITSLLDQIPEMFADTRETETVFVPVIQAGMEALKAAECAGKLFLFHTSLPIAEAPGKLKNRDDRKLINTDKEKTLFQPQTGAYQTLAKECVAQGCCVDLFLFPNQYVDVATLSVVPQLTGGSVYKYASFQVENDQERFLSDLRRDVQKVVGFDAVMRVRTSTGIRAVDFFGAFYMSNTTDVELAGLDGDKTVTVEFKHDDRLNEESGALLQCALLYTSCAGQRRLRIHNLALNCCTQLADLYRNCETDTLINYMAKFAYRGVLNSPVKAVRDTLITQCAQILACYRKNCASPSSAGQLILPECMKLLPVYLNCVLKSDVLQPGAEVTTDDRAYVRQLVTSMDVTETNVFFYPRLLPLTKSPVESTTEPPAVRASEERLSNGDIYLLENGLNLFLWVGASVQQGVVQSLFSVSSFSQITSGLSVLPVLDNPLSKKVRGLIDSLRAQRSRYMKLTVVKQEDKMEMLFKHFLVEDKSLSGGASYVDFLCHMHKEIRQLLSSpecifications
AAH18928 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
145.86kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0079 | |
SEC24C | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9632 | |
SEC24C (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNVNQSVPPVPPFGQPQPIYPGYHQSSYGGQSGSTAPAIPYGAYNGPVPGYQQTPPQGMSRAPPSSGAPPASTAQAPCGQAAYGQFGQGDVQNGPSSTVQMQRLPGSQSFGSPLAPVGNQPPVLQPYGPPPTSAQVATQLSGMQISGAVAPAPPSSGLGFGPPTSLASASGSFPNSGLYGSYPQGQAPPLSQAQGHPGIQTPQRSAPSQASSFTPPASGGPRLPSMTGPLLPGQSFGGPSVSQPNHVSSPPQALPPGTQMTGPLGPLPPMHSPQQPGYQPQQNGSFGPARGPQSNYGGPYPAAPTFGSQPGPPQPLPPKRLDPDAIPSPIQVIEDDRNNRGTEPFVTGVRGQVPPLVTTNFLVKDQGNASPRYIRCTSYNIPCTSDMAKQAQVPLAAVIKPLARLPPEEASPYVVDHGESGPLRCNRCKAYMCPFMQFIEGGRRFQCCFCSCINDVPPQYFQHLDHTGKRVDAYDRPELSLGSYEFLATVDYCKNNKFPSPPAFIFMIDVSYNAIRTGLVRLLCEELKSLLDFLPREGGAEESAIRVGFVTYNKVLHFYNVKSSLAQPQMMVVSDVADMFVPLLDGFLVNVNESRAVITSLLDQIPEMFADTRETETVFVPVIQAGMEALKAAECAGKLFLFHTSLPIAEAPGKLKNRDDRKLINTDKEKTLFQPQTGAYQTLAKECVAQGCCVDLFLFPNQYVDVATLSVVPQLTGGSVYKYASFQVENDQERFLSDLRRDVQKVVGFDAVMRVRTSTGIRAVDFFGAFYMSNTTDVELAGLDGDKTVTVEFKHDDRLNEESGALLQCALLYTSCAGQRRLRIHNLALNCCTQLADLYRNCETDTLINYMAKFAYRGVLNSPVKAVRDTLITQCAQILACYRKNCASPSSAGQLILPECMKLLPVYLNCVLKSDVLQPGAEVTTDDRAYVRQLVTSMDVTETNVFFYPRLLPLTKSPVESTTEPPAVRASEERLSNGDIYLLENGLNLFLWVGASVQQG | |
RUO | |
SEC24C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |