Learn More
Abnova™ Human SEC16B Full-length ORF (AAH09106.1, 1 a.a. - 625 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00089866-P01.10ug
Additional Details : Weight : 0.00010kg
Description
SEC16B is a mammalian homolog of S. cerevisiae Sec16 that is required for organization of transitional endoplasmic reticulum (ER) sites and protein export (Bhattacharyya and Glick, 2007 [PubMed 17192411]).[supplied by OMIM]
Sequence: MELWAPQRLPQTRGKATAPSKDPDRGFRRDGHHRPVPHSWHNGERFHQWQDNRGSPQPQQEPRADHQQQPHYASRPGDWHQPVSGVDYYEGGYRNQLYSRPGYENSYQSYQSPTMREEYAYGSYYYHGHPQWLQEERVPRQRSPYIWHEDYREQKYLDEHHYENQHSPFGTNSETHFQSNSRNPCKDSPASNSGQEWPGELFPGSLLAEAQKNKPSLASESNLLQQRESGLSSSSYELSQYIRDAPERDDPPASAAWSPVQAEDVSSAGPKAPMKFYIPHVPVSFGPGGQLVRVGPSSPTDGQAALVELHSMEVILNDSEEQEEMRSFSGPLIREDVHKVDIMTFCQQKAAQSCKSETLGSRDSALLWQLLVLLCRQNGSMVGSDIAELLMQDCKKLEKYKRQPPVANLINLTDEDWPVLSSGTPNLLTGEIPPSVETPAQIVEKFTRLLYYGRKKEALEWAMKNHLWGHALFLSSKMDPQTYSWVMSGFTSTLALNDPLQTLFQLMSGRIPQAATCCGEKQWGDWRPHLAVILSNQAGDPELYQRAIVAIGDTLAGKGLVEAAHFCYLMAHVPFGHYTVKTDHLVLLGSSHRYATWEKGNSKDIFQGTVLALVGFYGSSFHFLMSpecifications
AAH09106.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
97.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686C2486/FLJ23871/FLJ25761/FLJ33652/FLJ36620/LZTR2/PGPR-p117/RGPR/SEC16S | |
SEC16B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
89866 | |
SEC16B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MELWAPQRLPQTRGKATAPSKDPDRGFRRDGHHRPVPHSWHNGERFHQWQDNRGSPQPQQEPRADHQQQPHYASRPGDWHQPVSGVDYYEGGYRNQLYSRPGYENSYQSYQSPTMREEYAYGSYYYHGHPQWLQEERVPRQRSPYIWHEDYREQKYLDEHHYENQHSPFGTNSETHFQSNSRNPCKDSPASNSGQEWPGELFPGSLLAEAQKNKPSLASESNLLQQRESGLSSSSYELSQYIRDAPERDDPPASAAWSPVQAEDVSSAGPKAPMKFYIPHVPVSFGPGGQLVRVGPSSPTDGQAALVELHSMEVILNDSEEQEEMRSFSGPLIREDVHKVDIMTFCQQKAAQSCKSETLGSRDSALLWQLLVLLCRQNGSMVGSDIAELLMQDCKKLEKYKRQPPVANLINLTDEDWPVLSSGTPNLLTGEIPPSVETPAQIVEKFTRLLYYGRKKEALEWAMKNHLWGHALFLSSKMDPQTYSWVMSGFTSTLALNDPLQTLFQLMSGRIPQAATCCGEKQWGDWRPHLAVILSNQAGDPELYQRAIVAIGDTLAGKGLVEAAHFCYLMAHVPFGHYTVKTDHLVLLGSSHRYATWEKGNSKDIFQGTVLALVGFYGSSFHFLM | |
RUO | |
SEC16B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |