missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SDCCAG1 Partial ORF (NP_004704.2, 1010 a.a. - 1076 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009147-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120056

  • 5540.00 NOK / 25µg
Estimated Shipment: 22-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: KYKVKLTPGVQKKGKAAKTALNSFMHSKEATAREKDLFRSVKDTDLSRNIPGKVKVSAPNLLNVKRK
Specifications

Specifications

NP_004704.2
Liquid
9147
SDCCAG1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10051/NY-CO-1
SDCCAG1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.11kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KYKVKLTPGVQKKGKAAKTALNSFMHSKEATAREKDLFRSVKDTDLSRNIPGKVKVSAPNLLNVKRK
RUO
SDCCAG1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers