missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human SCF, AOF Recombinant Protein

Human SCF, AOF Recombinant Protein

Brand:  Gibco™ PHC6034

Additional Details : Weight : 0.05400kg

Product Code. 12003599

  • 3375.00 NOK / 10µg
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Human SCF, AOF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

SCF is a pleiotropic factor that acts in utero in germ cell and neural cell development, and hematopoiesis, all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human MO-7e cells
  • Proven compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture
10 ug
Human
Carrier-Free
proliferation of human TF-1 cells
10 μg
E. coli
Recombinant, Ligand
Other Cytokines
Gibco™
18.4 kDa
SCF AOF
Store in freezer (-5 to -30°C).
2 ng/ml
< 1 EU/ml
Wet Ice
Growth Factors & Receptors
Lyophilized
Immunology, Stem Cell Research, Cardiovascular Research, Neurobiology
98 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.