Learn More
Abnova™ Human SARS2 Full-length ORF (NP_060297.1, 1 a.a. - 518 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054938-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes the mitochondrial seryl-tRNA synthethase precursor, a member of the class II tRNA synthetase family. The mature enzyme catalyzes the ligation of Serine to tRNA(Ser) and participates in the biosynthesis of selenocysteinyl-tRNA(sec) in mitochondria. The enzyme contains an N-terminal tRNA binding domain and a core catalytic domain. It functions in a homodimeric form, which is stabilized by tRNA binding. This gene is regulated by a bidirectional promoter that also controls the expression of mitochondrial ribosomal protein S12. Both genes are within the critical interval for the autosomal dominant deafness locus DFNA4 and might be linked to this disease. Multiple transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MAASMARRLWPLLTRRGFRPRGGCISNDSPRRSFTTEKRNRNLLYEYAREGYSALPQLDIERFCACPEEAAHALELRKGELRSADLPAIISTWQELRQLQEQIRSLEEEKAAVTEAVRALLANQDSGEVQQDPKYQGLRARGREIRKELVHLYPREAQLEEQFYLQALKLPNQTHPDVPVGDESQARVLHMVGDKPVFSFQPRGHLEIGEKLDIIRQKRLSHVSGHRSYYLRGAGALLQHGLVNFTFNKLLRRGFTPMTVPDLLRGAVFEGCGMTPNANPSQIYNIDPARFKDLNLAGTAEVGLAGYFMDHTVAFRDLPVRMVCSSTCYRAETNTGQEPRGLYRVHHFTKVEMFGVTGPGLEQSSQLLEEFLSLQMEILTELGLHFRVLDMPTQELGLPAYRKFDIEAWMPGRGRFGEVTSASNCTDFQSRRLHIMFQTEAGELQFAHTVNATACAVPRLLIALLESNQQKDGSVLVPPALQSYLGTDRITAPTHVPLQYIGPNQPRKPGLPGQPAVSSpecifications
NP_060297.1 | |
Liquid | |
54938 | |
SARS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAASMARRLWPLLTRRGFRPRGGCISNDSPRRSFTTEKRNRNLLYEYAREGYSALPQLDIERFCACPEEAAHALELRKGELRSADLPAIISTWQELRQLQEQIRSLEEEKAAVTEAVRALLANQDSGEVQQDPKYQGLRARGREIRKELVHLYPREAQLEEQFYLQALKLPNQTHPDVPVGDESQARVLHMVGDKPVFSFQPRGHLEIGEKLDIIRQKRLSHVSGHRSYYLRGAGALLQHGLVNFTFNKLLRRGFTPMTVPDLLRGAVFEGCGMTPNANPSQIYNIDPARFKDLNLAGTAEVGLAGYFMDHTVAFRDLPVRMVCSSTCYRAETNTGQEPRGLYRVHHFTKVEMFGVTGPGLEQSSQLLEEFLSLQMEILTELGLHFRVLDMPTQELGLPAYRKFDIEAWMPGRGRFGEVTSASNCTDFQSRRLHIMFQTEAGELQFAHTVNATACAVPRLLIALLESNQQKDGSVLVPPALQSYLGTDRITAPTHVPLQYIGPNQPRKPGLPGQPAVS | |
RUO | |
SARS2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
84.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20450/SARS/SARSM/SERS/SYS/SerRSmt/mtSerRS | |
SARS2 | |
Yes | |
wheat germ expression system |