Learn More
Human SAMHD1 Antibody, R&D Systems™
GREENER_CHOICE
Product Code. 18109020
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
18109020 100 μg 100µg
1 options
Product Code. 18109020
Brand: R&D Systems MAB8120
Mouse Monoclonal Antibody
SAMHD1 Monoclonal specifically detects SAMHD1 in Human samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY
Specifications
Antigen SAMHD1
Applications Western Blot
Classification Monoclonal
Clone 883335
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y3Z3
Gene Alias AGS5, CHBL2, DCIP, DCIPMg11, Dendritic cell-derived IFNG-induced protein, EC 3.1.4.-, HDDC1, Monocyte protein 5, MOP5, MOP-5, MOP-5AGS5, SAM domain and HD domain 1, SAM domain and HD domain-containing protein 1, SBBI88
Gene Symbols SAMHD1
Host Species Mouse
Immunogen E. coli-derived recombinant human SAMHD1 His206-Arg339 Accession # Q9Y3Z3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25939
Test Specificity Detects human SAMHD1 in ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b