Learn More
Abnova™ Human RTN2 Full-length ORF (NP_996783.1, 1 a.a. - 472 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006253-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the family of reticulon encoding genes. Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MGQVLPVFAHCKEAPSTASSTPDSTEGGNDDSDFRELHTAREFSEEDEEETTSQDWGTPRELTFSYIAFDGVVGSGGRRDSTARRPRPQGRSVSEPRDQHPQPSLGDSLESIPSLSQSPEPGRRGDPDTAPPSERPLEDLRLRLDHLGWVARGTGSGEDSSTSSSTPLEDEEPQEPNRLETGEAGEELDLRLRLAQPSSPEVLTPQLSPGSGTPQAGTPSPSRSRDSNSGPEEPLLEEEEKQWGPLEREPVRGQCLDSTDQLEFTVEPRLLVADLLYWKDTRTSGVVFTGLMVSLLCLLHFSIVSVAAHLALLLLCGTISLRVYRKVLQAVHRGDGANPFQAYLDVDLTLTREQTERLSHQITSRVVSAATQLRHFFLVEDLVDSLKLALLFYILTFVGAIFNGLTLLILGVIGLFTIPLLYRQHQAQIDQYVGLVTNQLSHIKAKIRAKIPGTGALASAAAAVSGSKAKAESpecifications
NP_996783.1 | |
Liquid | |
6253 | |
RTN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGQVLPVFAHCKEAPSTASSTPDSTEGGNDDSDFRELHTAREFSEEDEEETTSQDWGTPRELTFSYIAFDGVVGSGGRRDSTARRPRPQGRSVSEPRDQHPQPSLGDSLESIPSLSQSPEPGRRGDPDTAPPSERPLEDLRLRLDHLGWVARGTGSGEDSSTSSSTPLEDEEPQEPNRLETGEAGEELDLRLRLAQPSSPEVLTPQLSPGSGTPQAGTPSPSRSRDSNSGPEEPLLEEEEKQWGPLEREPVRGQCLDSTDQLEFTVEPRLLVADLLYWKDTRTSGVVFTGLMVSLLCLLHFSIVSVAAHLALLLLCGTISLRVYRKVLQAVHRGDGANPFQAYLDVDLTLTREQTERLSHQITSRVVSAATQLRHFFLVEDLVDSLKLALLFYILTFVGAIFNGLTLLILGVIGLFTIPLLYRQHQAQIDQYVGLVTNQLSHIKAKIRAKIPGTGALASAAAAVSGSKAKAE | |
RUO | |
RTN2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NSP2/NSPL1 | |
RTN2 | |
Yes | |
wheat germ expression system |