Learn More
Abnova™ Human RP3-402G11.5 Full-length ORF (CAK54445.1, 1 a.a. - 509 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00083642-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. [provided by RefSeq]
Sequence: MYLGEVCTANGERWELQLKGAGPTPFSRQADGRKVLRSSIREFLCSEAMFHLGVPTTRAGACVTSESTVVRDVFYDGNPKYEQCTVVLRVASTFIRFGSFEIFKSADEHTGRAGPSVGRNDIRVQLLDYVISSFYPEIQAAHASDSVQRNAAFFREVTRRTARMVAEWQCVGFCHGVLNTDNMSILGLTIDYGPFGFLDRYDPDHVCNASDNTGRYAYSKQPEVCRWNLRKLAEALQPELPLELGEAILAEEFDAEFQRHYLQKMRRKLGLVQVELEEDGALVSKLLETMHLTGADFTNTFYLLSSFPVELESPGLAEFLARLMEQCASLEELRLAFRPQMDPRQLSMMLMLAQSNPQLFALMGTRAGIARELERVEQQSRLEQLSAAELQSRNQGHWADWLQAYRARLDKDLEGAGDAAAWQAEHVRVMHANNPKYVLRNYIAQNAIEAAERGDFSEVRRVLKLLETPYHCEAGAATDAEATEADGADGRQRSYSSKPPLWAAELCVTSpecifications
CAK54445.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.39kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC131879/SELO | |
RP3-402G11.5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
83642 | |
RP3-402G11.5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MYLGEVCTANGERWELQLKGAGPTPFSRQADGRKVLRSSIREFLCSEAMFHLGVPTTRAGACVTSESTVVRDVFYDGNPKYEQCTVVLRVASTFIRFGSFEIFKSADEHTGRAGPSVGRNDIRVQLLDYVISSFYPEIQAAHASDSVQRNAAFFREVTRRTARMVAEWQCVGFCHGVLNTDNMSILGLTIDYGPFGFLDRYDPDHVCNASDNTGRYAYSKQPEVCRWNLRKLAEALQPELPLELGEAILAEEFDAEFQRHYLQKMRRKLGLVQVELEEDGALVSKLLETMHLTGADFTNTFYLLSSFPVELESPGLAEFLARLMEQCASLEELRLAFRPQMDPRQLSMMLMLAQSNPQLFALMGTRAGIARELERVEQQSRLEQLSAAELQSRNQGHWADWLQAYRARLDKDLEGAGDAAAWQAEHVRVMHANNPKYVLRNYIAQNAIEAAERGDFSEVRRVLKLLETPYHCEAGAATDAEATEADGADGRQRSYSSKPPLWAAELCVT | |
RUO | |
RP3-402G11.5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |