Learn More
Abnova™ Human RNF19 Partial ORF (NP_056250, 739 a.a. - 837 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00025897-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded this gene contains two RING-finger motifs and an IBR (in between RING fingers) motif. This protein is an E3 ubiquintin ligase that is localized in Lewy bodies (LBs), a characteristic neuronal inclusion in Parkinson's disease (PD) brains. This protein interacts with UBE2L3/UBCH7 and UBE2E2/UBCH8, but not other ubiquitin-conjugating enzymes. This protein is found to bind and ubiquitylate synphilin 1 (SNCAIP), which is a interacting protein of alpha synuclein in neurons, and a major component of LB. Alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq]
Sequence: MKTSCSHGSSDYHTRFATVNILPEVENDRLENSPHQCSISVVTQTASCSEVSQLNHIAEEHGNNGIKPNVDLYFGDALKETNNNHSHQTMELKVAIQTESpecifications
NP_056250 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKTSCSHGSSDYHTRFATVNILPEVENDRLENSPHQCSISVVTQTASCSEVSQLNHIAEEHGNNGIKPNVDLYFGDALKETNNNHSHQTMELKVAIQTE | |
RUO | |
RNF19A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
25897 | |
RNF19 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp566B1346/DORFIN/RNF19 | |
RNF19A | |
Recombinant | |
wheat germ expression system |