missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF151 Partial ORF (NP_056484, 68 a.a. - 173 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00146310-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16142047

  • 5420.23 NOK / 25µg

Estimated Shipment: 17-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: HMNKLRKTIGRLEVKCKNADAGCIVTCPLAHRKGHQDSCPFELTACPNEGCTSQVPRGTLAEHRQHCQQGSQQRCPLGCGATLDPAERARHNCYRELHNAWSVRQE
Specifications

Specifications

NP_056484
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.4kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HMNKLRKTIGRLEVKCKNADAGCIVTCPLAHRKGHQDSCPFELTACPNEGCTSQVPRGTLAEHRQHCQQGSQQRCPLGCGATLDPAERARHNCYRELHNAWSVRQE
RUO
RNF151
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
146310
RNF151 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC129921
RNF151
Yes
Videos
SDS
Documents
Certificates