Learn More
Abnova™ Human RNF133 Full-length ORF (NP_631914.1, 1 a.a. - 376 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00168433-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene has no intron. [provided by RefSeq]
Sequence: MHLLKVGTWRNNTASSWLMKFSVLWLVSQNCCRASVVWMAYMNISFHVGNHVLSELGETGVFGRSSTLKRVAGVIVPPEGKIQNACNPNTIFSRSKYSETWLALIERGGCTFTQKIKVATEKGASGVIIYNVPGTGNQVFPMFHQAFEDVVVVMIGNLKGTEIFHLIKKGVLITAVVEVGRKHIIWMNHYLVSFVIVTTATLAYFIFYHIHRLCLARIQNRRWQRLTTDLQNTFGQLQLRVVKEGDEEINPNGDSCVICFERYKPNDIVRILTCKHFFHKNCIDPWILPHGTCPICKCDILKVLGIQVVVENGTEPLQVLMSNELPETLSPSEEETNNEVSPAGTSDKVIHVEENPTSQNNDIQPHSVVEDVHPSPSpecifications
NP_631914.1 | |
Liquid | |
168433 | |
RNF133 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MHLLKVGTWRNNTASSWLMKFSVLWLVSQNCCRASVVWMAYMNISFHVGNHVLSELGETGVFGRSSTLKRVAGVIVPPEGKIQNACNPNTIFSRSKYSETWLALIERGGCTFTQKIKVATEKGASGVIIYNVPGTGNQVFPMFHQAFEDVVVVMIGNLKGTEIFHLIKKGVLITAVVEVGRKHIIWMNHYLVSFVIVTTATLAYFIFYHIHRLCLARIQNRRWQRLTTDLQNTFGQLQLRVVKEGDEEINPNGDSCVICFERYKPNDIVRILTCKHFFHKNCIDPWILPHGTCPICKCDILKVLGIQVVVENGTEPLQVLMSNELPETLSPSEEETNNEVSPAGTSDKVIHVEENPTSQNNDIQPHSVVEDVHPSP | |
RUO | |
RNF133 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC27072 | |
RNF133 | |
Yes | |
wheat germ expression system |