Learn More
Abnova™ Human RNF13 Partial ORF (NP_009213, 51 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011342-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. The specific function of this gene has not yet been determined. Alternatively spliced transcript variants that encode the same protein have been reported. A pseudogene, which is also located on chromosome 3, has been defined for this gene. [provided by RefSeq]
Sequence: LPARFGYRLPAEGLKGFLINSKPENACEPIVPPPVKDNSSGTFIVLIRRLDCNFDIKVLNAQRAGYKAAIVHNVDSDDLISMGSNDIEVLKKIDIPSVFISpecifications
NP_009213 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LPARFGYRLPAEGLKGFLINSKPENACEPIVPPPVKDNSSGTFIVLIRRLDCNFDIKVLNAQRAGYKAAIVHNVDSDDLISMGSNDIEVLKKIDIPSVFI | |
RUO | |
RNF13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11342 | |
RNF13 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ93817/MGC13689/RZF | |
RNF13 | |
Recombinant | |
wheat germ expression system |