Learn More
Abnova™ Human RNF122 Full-length ORF (NP_079063.2, 1 a.a. - 155 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079845-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. [provided by RefSeq]
Sequence: MHPFQWCNGCFCGLGLVSTNKSCSMPPISFQDLPLNIYMVIFGTGIFVFMLSLIFCCYFISKLRNQAQSERYGYKEVVLKGDAKKLQLYGQTCAVCLEDFKGKDELGVLPCQHAFHRKCLVKWLEVRCVCPMCNKPIASPSEATQNIGILLDELVSpecifications
NP_079063.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12526/MGC126622 | |
RNF122 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
79845 | |
RNF122 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MHPFQWCNGCFCGLGLVSTNKSCSMPPISFQDLPLNIYMVIFGTGIFVFMLSLIFCCYFISKLRNQAQSERYGYKEVVLKGDAKKLQLYGQTCAVCLEDFKGKDELGVLPCQHAFHRKCLVKWLEVRCVCPMCNKPIASPSEATQNIGILLDELV | |
RUO | |
RNF122 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |