missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RKHD3 Partial ORF (NP_115622, 60 a.a. - 159 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084206-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140687

  • 3655.00 NOK / 10µg
Estimated Shipment: 23-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: RKKSVNMTECVPVPSSEHVAEIVGRQGCKIKALRAKTNTYIKTPVRGEEPVFVVTGRKEDVAMARREIISAAEHFSMIRASRNKNTALNGAVPGPPNLPG
Specifications

Specifications

NP_115622
Liquid
84206
RKHD3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434J0617/MEX-3B/MGC117199/RKHD3/RNF195
MEX3B
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RKKSVNMTECVPVPSSEHVAEIVGRQGCKIKALRAKTNTYIKTPVRGEEPVFVVTGRKEDVAMARREIISAAEHFSMIRASRNKNTALNGAVPGPPNLPG
RUO
MEX3B
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers