Learn More
Abnova™ Human RIPK4 Full-length ORF (BAB55102.1, 1 a.a. - 784 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054101-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a serine/threonine protein kinase that interacts with protein kinase C-delta. The encoded protein can also activate NFkappaB and is required for keratinocyte differentiation. This kinase undergoes autophosphorylation. [provided by RefSeq]
Sequence: MEGDGGTPWALALLRTFDAGEFTGWEKVGSGGFGQVYKVRHVHWKTWLAIKCSPSLHVDDRERMELLEEAKKMEMAKFRYILPVYGICREPVGLVMEYMETGSLEKLLASEPLPWDLRFRIIHETAVGMNFLHCMAPPLLHLDLKPANILLDAHYHVKISDFGLAKCNGLSHSHDLSMDGLFGTIAYLPPERIREKSRLSDTKHDVYSFAIVIWGVLTQKKPFADEKNILHIMVKVVKGHRPELPPVCRARPRACSHLIRLMQRCWQGDPRVRPTFQEITSETEDLCEKPDDEVKETAHDLDVKSPPEPRSEVVPARLKRASAPTFDNDYSLSELLSQLDSGVSQAVEGPEELSRSSSESKLPSSGSGKRLSGVSSVDSAFSSRGSLSLSFEREPSTSDLGTTDVQKKKLVDAIVSGDTSKLMKILQPQDVDLALDSGASLLHLAVEAGQEECAKWLLLNNANPNLSNRRGSTPLHMAVERRVRGVVERLLARKISVNAKDEDQWTALHFAAQNGDESSTRLLLEKNASVNEVDFEGLTPMHVACQHGQENIVRILLRRGVDVSLQGKDAWLPLHYAAWQGHLPIVKLLAKQPGVSVNAQTLDGRTPLHLAAQRGHYRVARILIDLCSDVNVCSLLAQTPLHVAAETGHTSTARLLLHRGAGKEAVTSDGYTALHLAARNGHLATVKLLVEEKADVLARGPLNQTALHLAAAHGHSEVVEELVSADVIDLFDEQGLSALHLAAQGRHAQTVETLLRHGAHINLQSLKFQGGHGPAATLLRRSKTSpecifications
BAB55102.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
112.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANKK2/ANKRD3/DIK/MGC129992/MGC129993/PKK/RIP4 | |
RIPK4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54101 | |
RIPK4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MEGDGGTPWALALLRTFDAGEFTGWEKVGSGGFGQVYKVRHVHWKTWLAIKCSPSLHVDDRERMELLEEAKKMEMAKFRYILPVYGICREPVGLVMEYMETGSLEKLLASEPLPWDLRFRIIHETAVGMNFLHCMAPPLLHLDLKPANILLDAHYHVKISDFGLAKCNGLSHSHDLSMDGLFGTIAYLPPERIREKSRLSDTKHDVYSFAIVIWGVLTQKKPFADEKNILHIMVKVVKGHRPELPPVCRARPRACSHLIRLMQRCWQGDPRVRPTFQEITSETEDLCEKPDDEVKETAHDLDVKSPPEPRSEVVPARLKRASAPTFDNDYSLSELLSQLDSGVSQAVEGPEELSRSSSESKLPSSGSGKRLSGVSSVDSAFSSRGSLSLSFEREPSTSDLGTTDVQKKKLVDAIVSGDTSKLMKILQPQDVDLALDSGASLLHLAVEAGQEECAKWLLLNNANPNLSNRRGSTPLHMAVERRVRGVVERLLARKISVNAKDEDQWTALHFAAQNGDESSTRLLLEKNASVNEVDFEGLTPMHVACQHGQENIVRILLRRGVDVSLQGKDAWLPLHYAAWQGHLPIVKLLAKQPGVSVNAQTLDGRTPLHLAAQRGHYRVARILIDLCSDVNVCSLLAQTPLHVAAETGHTSTARLLLHRGAGKEAVTSDGYTALHLAARNGHLATVKLLVEEKADVLARGPLNQTALHLAAAHGHSEVVEELVSADVIDLFDEQGLSALHLAAQGRHAQTVETLLRHGAHINLQSLKFQGGHGPAATLLRRSKT | |
RUO | |
RIPK4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |