Learn More
Abnova™ Human RHOBTB3 Partial ORF (NP_055714.2, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00022836-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
RHOBTB3 is a member of the evolutionarily conserved RHOBTB subfamily of Rho GTPases. For background information on RHOBTBs, see RHOBTB1 (MIM 607351).[supplied by OMIM]
Sequence: MSIHIVALGNEGDTFHQDNRPSGLIRTYLGRSPLVSGDESSLLLNAASTVARPVFTEYQASAFGNVKLVVHDCPVWDIFDSDWYTSRNLIGGADIIVIKSpecifications
NP_055714.2 | |
Liquid | |
22836 | |
RHOBTB3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0878 | |
RHOBTB3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSIHIVALGNEGDTFHQDNRPSGLIRTYLGRSPLVSGDESSLLLNAASTVARPVFTEYQASAFGNVKLVVHDCPVWDIFDSDWYTSRNLIGGADIIVIK | |
RUO | |
RHOBTB3 | |
Wheat Germ (in vitro) | |
GST |