missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RHEBL1 Partial ORF (AAH27482, 87 a.a. - 183 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00121268-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141637

  • 6270.00 NOK / 25µg
Estimated Shipment: 02-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: VTSLHSFQVIESLYQKLHEGHGKTRVPVVLVGNKADLSPEREVQAVEGKKLAESWGATFMESSARENQLTQGIFTKVIQEIARVENSYGQERRCHLM
Specifications

Specifications

AAH27482
Liquid
121268
RHEBL1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ25797/MGC34869/RHEBL1c
RHEBL1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.30kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VTSLHSFQVIESLYQKLHEGHGKTRVPVVLVGNKADLSPEREVQAVEGKKLAESWGATFMESSARENQLTQGIFTKVIQEIARVENSYGQERRCHLM
RUO
RHEBL1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers