Learn More
Abnova™ Human RGS5 Full-length ORF (NP_003608.1, 1 a.a. - 181 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008490-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The regulator of G protein signaling (RGS) proteins are signal transduction molecules that have structural homology to SST2 of Saccharomyces cerevisiae and EGL-10 of Caenorhabditis elegans. Multiple genes homologous to SST2 are present in higher eukaryotes. RGS proteins are involved in the regulation of heterotrimeric G proteins by acting as GTPase activators.[supplied by OMIM]
Sequence: MCKGLAALPHSCLERAKEIKIKLGILLQKPDSVGDLVIPYNEKPEKPAKTQKTSLDEALQWRDSLDKLLQNNYGLASFKSFLKSEFSEENLEFWIACEDYKKIKSPAKMAEKAKQIYEEFIQTEAPKEVNIDHFTKDITMKNLVEPSLSSFDMAQKRIHALMEKDSLPRFVRSEFYQELIKSpecifications
NP_003608.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MST092/MST106/MST129/MSTP032/MSTP092/MSTP106/MSTP129 | |
RGS5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8490 | |
RGS5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MCKGLAALPHSCLERAKEIKIKLGILLQKPDSVGDLVIPYNEKPEKPAKTQKTSLDEALQWRDSLDKLLQNNYGLASFKSFLKSEFSEENLEFWIACEDYKKIKSPAKMAEKAKQIYEEFIQTEAPKEVNIDHFTKDITMKNLVEPSLSSFDMAQKRIHALMEKDSLPRFVRSEFYQELIK | |
RUO | |
RGS5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |