missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RGS16 Partial ORF (NP_002919, 90 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006004-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134365

  • 4115.00 NOK / 10µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene belongs to the 'regulator of G protein signaling' family. It inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits. It also may play a role in regulating the kinetics of signaling in the phototransduction cascade. [provided by RefSeq]

Sequence: ENLEFWLACEEFKKIRSATKLASRAHQIFEEFICSEAPKEVNIDHETRELTRMNLQTATATCFDAAQGKTRTLMEKDSYPRFLKSPAYRD
Specifications

Specifications

NP_002919
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ENLEFWLACEEFKKIRSATKLASRAHQIFEEFICSEAPKEVNIDHETRELTRMNLQTATATCFDAAQGKTRTLMEKDSYPRFLKSPAYRD
RUO
RGS16
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6004
RGS16 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
A28-RGS14/A28-RGS14P/RGS-R
RGS16
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers