Learn More
Invitrogen™ Human RGPD5 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
RGPD5 Synthetic Peptide, PEP-0686, from Invitrogen.
The RANBP2-like and GRIP domain containing 5 protein (RGPD5) has high similarity to RANBP2, a large RAN-binding protein localized at the cytoplasmic side of the nuclear pore complex. The gene coding for RGPD5 is thought to have arisen from a gene duplication event of RANBP2 as these highly homologous genes are located close to each other at chromosome 2q11-q12. RGPD5 was identified as an HIV dependency factor (HDF), suggesting that RGPD5 may be an important drug target in HIV treatment. At least two isoforms of RGPD5 are known to exist, of which the shorter isoform is expressed primarily in testis, while the longer of the two is expressed at low levels in a number of somatic tissues.
Specifications
Specifications
| Gene ID (Entrez) | 84220 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q99666 |
| Common Name | Human RGPD5 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |