Learn More
Abnova™ Human RFX5 Full-length ORF (NP_000440.1, 1 a.a. - 616 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005993-P01.25ug
Additional Details : Weight : 0.00010kg
Description
A lack of MHC-II expression results in a severe immunodeficiency syndrome called MHC-II deficiency, or the bare lymphocyte syndrome (BLS; MIM 209920). At least 4 complementation groups have been identified in B-cell lines established from patients with BLS. The molecular defects in complementation groups B, C, and D all lead to a deficiency in RFX, a nuclear protein complex that binds to the X box of MHC-II promoters. The lack of RFX binding activity in complementation group C results from mutations in the RFX5 gene encoding the 75-kD subunit of RFX (Steimle et al., 1995). RFX5 is the fifth member of the growing family of DNA-binding proteins sharing a novel and highly characteristic DNA-binding domain called the RFX motif. Multiple alternatively spliced transcript variants have been found but the full-length natures of only two have been determined. [provided by RefSeq]
Sequence: MAEDEPDAKSPKTGGRAPPGGAEAGEPTTLLQRLRGTISKAVQNKVEGILQDVQKFSDNDKLYLYLQLPSGPTTGDKSSEPSTLSNEEYMYAYRWIRNHLEEHTDTCLPKQSVYDAYRKYCESLACCRPLSTANFGKIIREIFPDIKARRLGGRGQSKYCYSGIRRKTLVSMPPLPGLDLKGSESPEMGPEVTPAPRDELVEAACALTCDWAERILKRSFSSIVEVARFLLQQHLISARSAHAHVLKAMGLAEEDEHAPRERSSKPKNGLENPEGGAHKKPERLAQPPKDLEARTGAGPLARGERKKSVVESSAPGANNLQVNALVARLPLLLPRAPRSLIPPIPVSPPILAPRLSSGALKVATLPLSSRAGAPPAAVPIINMILPTVPALPGPGPGPGRAPPGGLTQPRGTENREVGIGGDQGPHDKGVKRTAEVPVSEASGQAPPAKAAKQDIEDTASDAKRKRGRPRKKSGGSGERNSTPLKSAAAMESAQSSRLPWETWGSGGEGNSAGGAERPGPMGEAEKGAVLAQGQGDGTVSKGGRGPGSQHTKEAEDKIPLVPSKVSVIKGSRSQKEAFPLAKGEVDTAPQGNKDLKEHVLQSSLSQEHKDPKATPPSpecifications
NP_000440.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RFX5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5993 | |
RFX5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEDEPDAKSPKTGGRAPPGGAEAGEPTTLLQRLRGTISKAVQNKVEGILQDVQKFSDNDKLYLYLQLPSGPTTGDKSSEPSTLSNEEYMYAYRWIRNHLEEHTDTCLPKQSVYDAYRKYCESLACCRPLSTANFGKIIREIFPDIKARRLGGRGQSKYCYSGIRRKTLVSMPPLPGLDLKGSESPEMGPEVTPAPRDELVEAACALTCDWAERILKRSFSSIVEVARFLLQQHLISARSAHAHVLKAMGLAEEDEHAPRERSSKPKNGLENPEGGAHKKPERLAQPPKDLEARTGAGPLARGERKKSVVESSAPGANNLQVNALVARLPLLLPRAPRSLIPPIPVSPPILAPRLSSGALKVATLPLSSRAGAPPAAVPIINMILPTVPALPGPGPGPGRAPPGGLTQPRGTENREVGIGGDQGPHDKGVKRTAEVPVSEASGQAPPAKAAKQDIEDTASDAKRKRGRPRKKSGGSGERNSTPLKSAAAMESAQSSRLPWETWGSGGEGNSAGGAERPGPMGEAEKGAVLAQGQGDGTVSKGGRGPGSQHTKEAEDKIPLVPSKVSVIKGSRSQKEAFPLAKGEVDTAPQGNKDLKEHVLQSSLSQEHKDPKATPP | |
RUO | |
RFX5 | |
Recombinant | |
wheat germ expression system |