Learn More
STEMCELL Technologies Human Recombinant MCP-1 (CCL2)
Click to view available options
Quantity:
20 μg
5 μg
Unit Size:
20µg
5µg
Description
- Monocyte chemotactic protein-1 (MCP-1), also known as CCL2, is a member of the CC family of chemokines
- The protein is primarily induced by platelet-derived growth factor (PDGF) gene (Cochran et al.) The biological effects of MCP-1 are mediated via the specific G-protein-coupled receptor CCR2 which in turn activates signal transduction pathways leading to monocyte transmigration
- Migration of monocytes from the bloodstream across the vascular endothelium is required for routine immunological surveillance of tissues, as well as other immunomodulatory effects
- MCP-1 is produced by a variety of cell types, including fibroblasts and endothelial, epithelial, smooth muscle, mesangial, astrocytic, monocytic, and microglial cells, which are important for antiviral responses in the peripheral circulations and in tissues
Specifications
Specifications
| Name | DIA, Differentiation-inducing factor, HILDA, Human interleukin in DA cells, Leukemia inhibitory factor, MCAF, Melanoma-derived LPL inhibitor, MLPLI, Monocyte chemoattractant protein 1 |
| Quantity | 5 μg |
| Cell Type | Macrophages; Monocytes; Myeloid Cells; NK Cells; T Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.