missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant IL-7, ACF

Product Code. 17109811
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
This item is not returnable. View return policy

Product Code. 17109811

Brand: STEMCELL Technologies 78196

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin 7, animal component-free

  • Interleukin 7 (IL-7) is a member of the type I cytokine family that is critical for T and B cell development and survival
  • It is produced by non-hematopoietic cells in the thymus, lymphoid organs, and by bone marrow stromal cells (Lundström et al.) IL-7 binds to a receptor (IL-7R) composed of common gamma chain and IL-7Ra (CD127) and signals through the JAK/STAT and PI3K pathways
  • IL-7 regulates the survival of naïve and memory CD4+ and CD8+ T cells, γδ T cells, NK T cells, innate lymphoid cells, and regulatory T cells (Carrette & Surh)
  • Although a deficiency in IL-7R still permits the generation of normal numbers of peripheral B cells in humans, stimulation of human B cell precursors with IL-7 could promote STAT5-dependent proliferation and survival in vitro (Clark et al.; Corfe & Paige)
  • This product is animal component-free

Specifications

Name Interleukin-7, LP-1, Lymphopoietin 1, Pre-B cell factor
Quantity 10 μg
Cell Type Hematopoietic Stem and Progenitor Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.