Learn More
Abnova™ Human RBMY1A1 Partial ORF (NP_001007527.1, 257 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005940-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein containing an RNA-binding motif in the N-terminus and four SRGY (serine, arginine, glycine, tyrosine) boxes in the C-terminus. Multiple copies of this gene are found in the AZFb azoospermia factor region of chromosome Y and the encoded protein is thought to be involved in spermatogenesis. Most copies of this locus are pseudogenes, although six highly similar copies have full-length ORFs and are considered functional. Four functional copies of this gene are found within inverted repeat IR2; two functional copies of this gene are found in palindrome P3, along with two copies of PTPN13-like, Y-linked. [provided by RefSeq]
Sequence: YGTSHGAPPARGPRMSYGGSTCHAYSNTRDRYGRSWESYSSCGDFHYCDREHVCRKDQRNPPSLGRVLPDPREACGSSSYVASIVDGGESRSEKGDSSRYSpecifications
NP_001007527.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YGTSHGAPPARGPRMSYGGSTCHAYSNTRDRYGRSWESYSSCGDFHYCDREHVCRKDQRNPPSLGRVLPDPREACGSSSYVASIVDGGESRSEKGDSSRY | |
RUO | |
RBMY1A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5940 | |
RBMY1A1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC181956/RBM1/RBM2/RBMY/YRRM1/YRRM2 | |
RBMY1A1 | |
Recombinant | |
wheat germ expression system |