missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RBM8A Full-length ORF (AAH17088, 1 a.a. - 174 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009939-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194242

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein with a conserved RNA-binding motif. The protein is found predominantly in the nucleus, although it is also present in the cytoplasm. It is preferentially associated with mRNAs produced by splicing, including both nuclear mRNAs and newly exported cytoplasmic mRNAs. It is thought that the protein remains associated with spliced mRNAs as a tag to indicate where introns had been present, thus coupling pre- and post-mRNA splicing events. Previously, it was thought that two genes encode this protein, RBM8A and RBM8B; it is now thought that the RBM8B locus is a pseudogene. Two alternative start codons result in two forms of the protein, and this gene also uses multiple polyadenylation sites. [provided by RefSeq]

Sequence: MADVLDLHEAGGEDFAMDEDGDESIHKLKEKAKKRKGRGFGSEEGSRARMREDYDSVEQDGDEPGPQRSVEGWILSVTGVHEEATEEDIHDKFAEYGEIKNIHLNLDRRTGYLKGYTLVEYETYKEAQAAMEGLNGQDLMGQPISVDWCFVRGPPKGKRRGGRRRSRSPDRRRR
Specifications

Specifications

AAH17088
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.88kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BOV-1A/BOV-1B/BOV-1C/MDS014/RBM8/RBM8B/Y14/ZNRP/ZRNP1
RBM8A
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9939
RBM8A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MADVLDLHEAGGEDFAMDEDGDESIHKLKEKAKKRKGRGFGSEEGSRARMREDYDSVEQDGDEPGPQRSVEGWILSVTGVHEEATEEDIHDKFAEYGEIKNIHLNLDRRTGYLKGYTLVEYETYKEAQAAMEGLNGQDLMGQPISVDWCFVRGPPKGKRRGGRRRSRSPDRRRR
RUO
RBM8A
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers