missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RASGRP4 Partial ORF (NP_733748, 574 a.a. - 673 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00115727-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141497

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the Ras guanyl nucleotide-releasing protein (RasGRP) family of Ras guanine nucleotide exchange factors. It contains a Ras exchange motif, a diacylglycerol-binding domain, and two calcium-binding EF hands. This protein was shown to activate H-Ras in a cation-dependent manner in vitro. Expression of this protein in myeloid cell lines was found to be correlated with elevated level of activated RAS protein, and the RAS activation can be greatly enhanced by phorbol ester treatment, which suggested a role of this protein in diacylglycerol regulated cell signaling pathways. Studies of a mast cell leukemia cell line expressing substantial amounts of abnormal transcripts of this gene indicated that this gene may play an important role in the final stages of mast cell development. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: CGLCCHKHCRDQVKVECKKRPGAKGDAGPPGAPVPSTPAPHASCGSEENHSYTLSLEPETGCQLRHAWTQTESPHPSWETDTVPCPVMDPPSTASSKLDS
Specifications

Specifications

NP_733748
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CGLCCHKHCRDQVKVECKKRPGAKGDAGPPGAPVPSTPAPHASCGSEENHSYTLSLEPETGCQLRHAWTQTESPHPSWETDTVPCPVMDPPSTASSKLDS
RUO
RASGRP4
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
115727
RASGRP4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RASGRP4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers